3-(tert-Butyldimethylsilyloxy)-5-(methoxycarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-((tert-butyldimethylsilyl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-5979-1G PN-5979-250M
COBBPN-5979-1GEA 0 GBP
COBBPN-5979-1G COBBPN-5979-250M
3-(tert-Butyldimethylsilyloxy)-5-(methoxycarbonyl)phenylboronic acid pinacol ester
3-(tert-Butyldimethylsilyloxy)-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₂₀H₃₃BO₅Si
MW: 392.37 g/mol
MDL Number: MFCD12546599
CAS Number: 1218789-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR