N(α)-Benzyloxycarbonyl-L-lysine

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH

4001304.0005 4001304.0025
BACE4001304.0005EA 0 GBP
BACE4001304.0005 BACE4001304.0025
N(α)-Benzyloxycarbonyl-L-lysine
N(α)-Benzyloxycarbonyl-L-lysine
Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
MDL Number: MFCD00038204
CAS Number: 2212-75-1
EINECS: 218-662-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR