N-Boc-L-valine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Boc-Val-OH, BOC-L-Valine

15528-100G 15528-25G
SIAL15528-100GEA 103 GBP
SIAL15528-100G SIAL15528-25G
N-Boc-L-valine, Sigma-Aldrich®
N-Boc-L-valine
Formula: C₁₀H₁₉NO₄
MW: 217.3 g/mol
MDL Number: MFCD00065605
CAS Number: 13734-41-3
EINECS: 237-307-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR