N-Boc-D-valine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Boc-D-Val-OH
AS22209G100
AS22209G25
ACTIAS22209G100EA
0
GBP
ACTIAS22209G100
ACTIAS22209G25
N-Boc-D-valine
N-Boc-D-valine
Formula:
C₁₀H₁₉NO₄ MW: 217.3 g/mol |
MDL Number:
MFCD00038282 CAS Number: 22838-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...