Ethyl picolinoylacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1094.1 023365-1G 023365-5G
CHMPCFF1094.1EA 0 GBP
CHMPCFF1094.1 FLUO023365-1G FLUO023365-5G
Ethyl picolinoylacetate
Ethyl picolinoylacetate
Formula: C₁₀H₁₁NO₃
MDL Number: MFCD00094022
CAS Number: 26510-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR