(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: [(5-Sulphanyl-1,3,4-thiadiazol-2-yl)sulphanyl]acetic acid

009879-1G 009879-5G
FLUO009879-1GEA 0 GBP
FLUO009879-1G FLUO009879-5G
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
Formula: C₄H₄N₂O₂S₃
MW: 208.28 g/mol
MDL Number: MFCD00129970
CAS Number: 53723-88-9
EINECS: 258-728-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR