4-(Methanesulphonylamino)benzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(Methylsulphonylamino)benzonitrile
AS20867G25
AS20867G5
ACTIAS20867G25EA
354
GBP
ACTIAS20867G25
ACTIAS20867G5
4-(Methanesulphonylamino)benzonitrile
4-(Methanesulphonylamino)benzonitrile
Formula:
C₈H₈N₂O₂S MW: 196.23 g/mol |
MDL Number:
MFCD00204724 CAS Number: 36268-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...