3-(4-Cyanophenoxy)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22929G1
AS22929G5
ACTIAS22929G1EA
0
GBP
ACTIAS22929G1
ACTIAS22929G5
3-(4-Cyanophenoxy)propionic acid
3-(4-Cyanophenoxy)propionic acid
Formula:
C₁₀H₉NO₃ |
MDL Number:
MFCD09043034 CAS Number: 58228-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...