3-(3-Cyanophenoxy)propionic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22930G1
AS22930G5
ACTIAS22930G1EA
0
GBP
ACTIAS22930G1
ACTIAS22930G5
3-(3-Cyanophenoxy)propionic acid
3-(3-Cyanophenoxy)propionic acid
Formula:
C₁₀H₉NO₃ |
MDL Number:
MFCD09812167 CAS Number: 210962-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...