4-(5-Oxazolyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23777G1 AS23777G5
ACTIAS23777G1EA 0 GBP
ACTIAS23777G1 ACTIAS23777G5
4-(5-Oxazolyl)benzonitrile
4-(5-Oxazolyl)benzonitrile
Formula: C₁₀H₆N₂O
MDL Number: MFCD00084987
CAS Number: 87150-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR