2-(Methylthio)thiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009622-25G
009622-5G
FLUO009622-25GEA
0
GBP
FLUO009622-25G
FLUO009622-5G
2-(Methylthio)thiazole
2-(Methylthio)thiazole
Formula:
C₄H₅NS₂ MW: 131.21 g/mol |
MDL Number:
MFCD00858981 CAS Number: 5053-24-7 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...