2-(Methylthio)thiazole ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B25137.06 B25137.14 B25137.22
B25137.06EA 0 GBP
B25137.06 B25137.14 B25137.22
2-(Methylthio)thiazole ≥98%
2-(Methylthio)thiazole

Purity: 98% CAS No.: 5053-24-7 Molecular Formula: C4H5NS2 Formula Weight: 131.21 Flash Point: 90°(195°F) Refractive Index: 1.6080 Odour: Characteristic Boiling Point: 205-207° MDL No.: MFCD00858981

Formula: C₄H₅NS₂
MW: 131.21 g/mol
Boiling Pt: 205…207 °C
Density: 1.271
Flash Pt: 90 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00858981
CAS Number: 5053-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR