Tributyl(4-chlorophenyl)stannane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
721158-1G
721158-5G
SIAL721158-1GEA
90
GBP
SIAL721158-1G
SIAL721158-5G
Tributyl(4-chlorophenyl)stannane, Sigma-Aldrich®
Tributyl(4-chlorophenyl)stannane
Formula:
C₁₈H₃₁ClSn |
CAS Number:
17151-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...