cis-1-Ethoxy-2-(tributylstannyl)ethene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51184G1 AS51184G25 AS51184G5
ACTIAS51184G1EA 0 GBP
ACTIAS51184G1 ACTIAS51184G25 ACTIAS51184G5
cis-1-Ethoxy-2-(tributylstannyl)ethene
cis-1-Ethoxy-2-(tributylstannyl)ethene
Formula: C₁₆H₃₄OSn
MDL Number: MFCD07787391
CAS Number: 64724-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR