1-Ethoxy-1-(tributylstannyl)ethene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin
AS51001G25
AS51001G5
ACTIAS51001G25EA
59
GBP
ACTIAS51001G25
ACTIAS51001G5
1-Ethoxy-1-(tributylstannyl)ethene
1-Ethoxy-1-(tributylstannyl)ethene
Formula:
C₁₆H₃₄OSn MW: 361.15 g/mol |
MDL Number:
MFCD00010240 CAS Number: 97674-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...