2-Benzyloxy-5-pyrimidineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5859-1G
BB-5859-250M
COBBBB-5859-1GEA
0
GBP
COBBBB-5859-1G
COBBBB-5859-250M
2-Benzyloxy-5-pyrimidineboronic acid
2-Benzyloxy-5-pyrimidineboronic acid
Formula:
C₁₁H₁₁BN₂O₃ |
MDL Number:
MFCD07375141 CAS Number: 1217500-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...