Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1071.1 005213-5G 005213-100G 005213-25G
CHMPCFF1071.1EA 0 GBP
CHMPCFF1071.1 FLUO005213-5G FLUO005213-100G FLUO005213-25G
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula: C₈H₉ClN₂O₂S
MDL Number: MFCD00006085
CAS Number: 5909-24-0
EINECS: 227-619-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR