2-(Methylthio)-5-pyrimidineboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid
016212-1G
016212-5G
FLUO016212-1GEA
0
GBP
FLUO016212-1G
FLUO016212-5G
2-(Methylthio)-5-pyrimidineboronic acid
2-(Methylthio)-5-pyrimidineboronic acid
Formula:
C₅H₇BN₂O₂S |
MDL Number:
MFCD07375142 CAS Number: 348098-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...