2-(Methylamino)-5-pyrimidineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methylaminopyrimidine-5-boronic acid, pinacol ester, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5920-1G PN-5920-250M
COBBPN-5920-1GEA 0 GBP
COBBPN-5920-1G COBBPN-5920-250M
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester
2-(Methylamino)-5-pyrimidineboronic acid pinacol ester
Formula: C₁₁H₁₈BN₃O₂
MDL Number: MFCD11878345
CAS Number: 904326-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR