1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5118-5G BB-5118-1G
COBBBB-5118-5GEA 0 GBP
COBBBB-5118-5G COBBBB-5118-1G
1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid
1-(3-Chlorophenyl)-1H-pyrazole-4-boronic acid
Formula: C₉H₈BClN₂O₂
MDL Number: MFCD09878353
CAS Number: 1072945-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR