1-(4-(4-Bromo-1H-pyrazol-1-yl)phenylsulphonyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1115-5G OT-1115-1G
COBBOT-1115-5GEA 0 GBP
COBBOT-1115-5G COBBOT-1115-1G
1-(4-(4-Bromo-1H-pyrazol-1-yl)phenylsulphonyl)piperidine
1-(4-(4-Bromo-1H-pyrazol-1-yl)phenylsulphonyl)piperidine
Formula: C₁₄H₁₆BrN₃O₂S
MDL Number: MFCD13195747
CAS Number: 1199773-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR