1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0843-5G
OT-0843-1G
COBBOT-0843-5GEA
0
GBP
COBBOT-0843-5G
COBBOT-0843-1G
1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole
1-(2-(4-Bromophenoxy)ethyl)-1H-pyrazole
Formula:
C₁₁H₁₁BrN₂O |
MDL Number:
MFCD11855961 CAS Number: 1150271-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...