1,2-Dimethylimidazole-4-sulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,2-Dimethyl-1H-imidazole-4-sulphonyl chloride

CFF227.1 CFF227.5 017280-1G 017280-5G
CHMPCFF227.1EA 0 GBP
CHMPCFF227.1 CHMPCFF227.5 FLUO017280-1G FLUO017280-5G
1,2-Dimethylimidazole-4-sulphonyl chloride
1,2-Dimethylimidazole-4-sulphonyl chloride
Formula: C₅H₇ClN₂O₂S
MDL Number: MFCD01313780
CAS Number: 137049-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR