(1-Methyl-1H-imidazol-2-yl)methanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-2-imidazolemethanol

AS20763G100 AS20763G25 AS20763G5
ACTIAS20763G100EA 113 GBP
ACTIAS20763G100 ACTIAS20763G25 ACTIAS20763G5
(1-Methyl-1H-imidazol-2-yl)methanol
(1-Methyl-1H-imidazol-2-yl)methanol
Formula: C₅H₈N₂O
MW: 112.13 g/mol
MDL Number: MFCD00964673
CAS Number: 17334-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR