1-Methylimidazole-4-sulphonamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF312.0.25 CFF312.0.5 CFF312.1 017352-250MG 017352-500MG 017352-1G
CHMPCFF312.0.25EA 0 GBP
CHMPCFF312.0.25 CHMPCFF312.0.5 CHMPCFF312.1 FLUO017352-250MG FLUO017352-500MG FLUO017352-1G
1-Methylimidazole-4-sulphonamide
1-Methylimidazole-4-sulphonamide
Formula: C₄H₇N₃O₂S
MDL Number: MFCD02091382
CAS Number: 111124-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR