1-(Ethoxymethyl)imidazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1021-5G
HI-1021-1G
COBBHI-1021-5GEA
0
GBP
COBBHI-1021-5G
COBBHI-1021-1G
1-(Ethoxymethyl)imidazole
1-(Ethoxymethyl)imidazole
Formula:
C₆H₁₀N₂O |
MDL Number:
MFCD09878401 CAS Number: 67319-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...