1,4-Benzodioxane-2-carbonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

017521-1G 017521-5G
FLUO017521-1GEA 0 GBP
FLUO017521-1G FLUO017521-5G
1,4-Benzodioxane-2-carbonyl chloride
1,4-Benzodioxane-2-carbonyl chloride
Formula: C₉H₇ClO₃
MDL Number: MFCD02180342
CAS Number: 3663-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR