7-(Trifluoromethyl)isatin

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0122-5G IN-0122-1G
COBBIN-0122-5GEA 0 GBP
COBBIN-0122-5G COBBIN-0122-1G
7-(Trifluoromethyl)isatin
7-(Trifluoromethyl)isatin
Formula: C₉H₄F₃NO₂
MDL Number: MFCD01248408
CAS Number: 391-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR