7-(Trifluoromethyl)isatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21010G25 AS21010G5
ACTIAS21010G25EA 0 GBP
ACTIAS21010G25 ACTIAS21010G5
7-(Trifluoromethyl)isatin
7-(Trifluoromethyl)isatin
Formula: C₉H₄F₃NO₂
MDL Number: MFCD01248408
CAS Number: 391-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR