7-(Trifluoromethoxy)isatin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS35690G1 AS35690G5
ACTIAS35690G1EA 0 GBP
ACTIAS35690G1 ACTIAS35690G5
7-(Trifluoromethoxy)isatin
7-(Trifluoromethoxy)isatin
Formula: C₉H₄F₃NO₃
MDL Number: MFCD06804535
CAS Number: 149125-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR