1-Methyl-1H-benzimidazole-6-boronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5116-1G
BB-5116-250M
COBBBB-5116-1GEA
0
GBP
COBBBB-5116-1G
COBBBB-5116-250M
1-Methyl-1H-benzimidazole-6-boronic acid
1-Methyl-1H-benzimidazole-6-boronic acid
Formula:
C₈H₉BN₂O₂ |
MDL Number:
MFCD09972099 CAS Number: 1072945-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...