1H-Benzimidazole-5-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzimidazole, Benzimidazole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole

AS12189G1 AS12189G5
ACTIAS12189G1EA 0 GBP
ACTIAS12189G1 ACTIAS12189G5
1H-Benzimidazole-5-boronic acid pinacol ester
1H-Benzimidazole-5-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MDL Number: MFCD11054041
CAS Number: 1007206-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR