1-((4-Bromophenyl)sulphonyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0088-5G OT-0088-1G
COBBOT-0088-5GEA 0 GBP
COBBOT-0088-5G COBBOT-0088-1G
1-((4-Bromophenyl)sulphonyl)piperidine
1-((4-Bromophenyl)sulphonyl)piperidine
Formula: C₁₁H₁₄BrNO₂S
MW: 304.2034 g/mol
MDL Number: MFCD00194753
CAS Number: 834-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR