1-((4-Bromophenyl)sulphonyl)piperidin-4-one

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0502-5G OT-0502-1G
COBBOT-0502-5GEA 0 GBP
COBBOT-0502-5G COBBOT-0502-1G
1-((4-Bromophenyl)sulphonyl)piperidin-4-one
1-((4-Bromophenyl)sulphonyl)piperidin-4-one
Formula: C₁₁H₁₂BrNO₃S
MDL Number: MFCD09258649
CAS Number: 929000-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR