1-Boc-4-(cyanomethyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6324-5G HC-6324-1G
COBBHC-6324-5GEA 0 GBP
COBBHC-6324-5G COBBHC-6324-1G
1-Boc-4-(cyanomethyl)piperidine
1-Boc-4-(cyanomethyl)piperidine
Formula: C₁₂H₂₀N₂O₂
MDL Number: MFCD09801019
CAS Number: 256411-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR