1-(4-Chlorobenzoyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4519-5G CA-4519-25G CA-4519-1G
COBBCA-4519-5GEA 0 GBP
COBBCA-4519-5G COBBCA-4519-25G COBBCA-4519-1G
1-(4-Chlorobenzoyl)pyrrolidine
1-(4-Chlorobenzoyl)pyrrolidine
Formula: C₁₁H₁₂ClNO
MDL Number: MFCD00587049
CAS Number: 19202-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR