1-(4-Bromo-2-chlorobenzoyl)piperidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4738-1G CA-4738-5G
COBBCA-4738-1GEA 0 GBP
COBBCA-4738-1G COBBCA-4738-5G
1-(4-Bromo-2-chlorobenzoyl)piperidine
1-(4-Bromo-2-chlorobenzoyl)piperidine
Formula: C₁₂H₁₃BrClNO
MDL Number: MFCD12756428
CAS Number: 1187385-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR