(S)-1-Boc-3-(hydroxymethyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS3142G1 AS3142G25 AS3142G5
ACTIAS3142G1EA 27 GBP
ACTIAS3142G1 ACTIAS3142G25 ACTIAS3142G5
(S)-1-Boc-3-(hydroxymethyl)piperazine
(S)-1-Boc-3-(hydroxymethyl)piperazine
Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD05863889
CAS Number: 314741-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR