(R)-1-Boc-3-(Hydroxymethyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-tert-Butyl 3-(hydroxymethyl)piperazine-1-carboxylate, (3R)-3-(Hydroxymethyl)piperazine, N1-BOC protected

AS3141G5 AS3141G25
ACTIAS3141G5EA 0 GBP
ACTIAS3141G5 ACTIAS3141G25
(R)-1-Boc-3-(Hydroxymethyl)piperazine
(R)-1-Boc-3-(hydroxymethyl)piperazine
Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD05863888
CAS Number: 278788-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR