1-Boc-3-isobutylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12163G5 AS12163G1
ACTIAS12163G5EA 0 GBP
ACTIAS12163G5 ACTIAS12163G1
1-Boc-3-isobutylpiperazine
1-Boc-3-isobutylpiperazine
Formula: C₁₃H₂₆N₂O₂
MDL Number: MFCD08273962
CAS Number: 78551-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR