1-Boc-2-methylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methylpiperazine, N1-BOC protected, tert-Butyl 2-methylpiperazine-1-carboxylate

AS21746G25 AS21746G5
ACTIAS21746G25EA 0 GBP
ACTIAS21746G25 ACTIAS21746G5
1-Boc-2-methylpiperazine
1-Boc-2-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
MDL Number: MFCD02179168
CAS Number: 120737-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR