(S)-1-Boc-3-propylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS3127G5 AS3127G1
ACTIAS3127G5EA 0 GBP
ACTIAS3127G5 ACTIAS3127G1
(S)-1-Boc-3-propylpiperazine
(S)-1-Boc-3-propylpiperazine
Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD07772106
CAS Number: 928025-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR