(S)-(-)-1-Boc-3-methylpiperazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AM-1755-5G
AM-1755-25G
AM-1755-1G
COBBAM-1755-5GEA
0
GBP
COBBAM-1755-5G
COBBAM-1755-25G
COBBAM-1755-1G
(S)-(-)-1-Boc-3-methylpiperazine
(S)-(-)-1-Boc-3-methylpiperazine
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD02683204 CAS Number: 147081-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...