(S)-(-)-1-Boc-3-methylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12154G25 AS12154G5
ACTIAS12154G25EA 0 GBP
ACTIAS12154G25 ACTIAS12154G5
(S)-(-)-1-Boc-3-methylpiperazine
(S)-(-)-1-Boc-3-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD02683204
CAS Number: 147081-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR