(S)-1-Boc-2-methylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS3110G1 AS3110G25 AS3110G5
ACTIAS3110G1EA 0 GBP
ACTIAS3110G1 ACTIAS3110G25 ACTIAS3110G5
(S)-1-Boc-2-methylpiperazine
(S)-1-Boc-2-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01862121
CAS Number: 169447-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR