(R)-1-Boc-3-Isobutylpiperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS3140G1 AS3140G5
ACTIAS3140G1EA 0 GBP
ACTIAS3140G1 ACTIAS3140G5
(R)-1-Boc-3-Isobutylpiperazine
(R)-1-Boc-3-isobutylpiperazine
Formula: C₁₃H₂₆N₂O₂
MDL Number: MFCD07772109
CAS Number: 928025-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR