4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)boronic acid
BB-8197-1G
BB-8197-250M
COBBBB-8197-1GEA
0
GBP
COBBBB-8197-1G
COBBBB-8197-250M
4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid
4-(4-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid
Formula:
C₁₅H₁₆BClO₃ MW: 290.5500 g/mol |
MDL Number:
MFCD06411357 CAS Number: 849062-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...