3,5-Dimethyl-4-(3',5'-dimethoxybenzyloxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8205-1G BB-8205-250M
COBBBB-8205-1GEA 0 GBP
COBBBB-8205-1G COBBBB-8205-250M
3,5-Dimethyl-4-(3',5'-dimethoxybenzyloxy)phenylboronic acid
3,5-Dimethyl-4-(3',5'-dimethoxybenzyloxy)phenylboronic acid
Formula: C₁₇H₂₁BO₅
MDL Number: MFCD08276774
CAS Number: 1072951-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR