4-(4-Fluorophenylsulphonyl)morpholine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0970-5G
OT-0970-1G
COBBOT-0970-5GEA
0
GBP
COBBOT-0970-5G
COBBOT-0970-1G
4-(4-Fluorophenylsulphonyl)morpholine
4-(4-Fluorophenylsulphonyl)morpholine
Formula:
C₁₀H₁₂FNO₃S |
MDL Number:
MFCD00194713 CAS Number: 383-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...