1-Boc-4-cyanopiperidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20379G25 AS20379G1 AS20379G5
ACTIAS20379G25EA 0 GBP
ACTIAS20379G25 ACTIAS20379G1 ACTIAS20379G5
1-Boc-4-cyanopiperidine
1-Boc-4-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
MDL Number: MFCD01861223
CAS Number: 91419-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR